missing translation for 'onlineSavingsMsg'
Learn More

Ethylurea, 98%

Catalog No. AAB2404036
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2404036 500 g
AAB2404014 25 g
AAB2404022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2404036 Supplier Thermo Scientific Chemicals Supplier No. B2404036
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 625-52-5 | C3H8N2O | 88.11 g/mol

N-Ethyl urea, is a versatile building block, that can be used in organic synthesis of various pharmaceutical and biologically active compounds, as nitrogen and carbonyl source.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Ethyl urea, is a versatile building block, that can be used in organic synthesis of various pharmaceutical and biologically active compounds, as nitrogen and carbonyl source.

Solubility
Soluble in methanol (25 mg/ml), and water (100 mg/ml).

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 625-52-5
Molecular Formula C3H8N2O
Molecular Weight (g/mol) 88.11
MDL Number MFCD00007953
InChI Key RYECOJGRJDOGPP-UHFFFAOYSA-N
Synonym 1-ethylurea, n-ethylurea, urea, ethyl, urea, 1-ethyl, polyurethane, urea, n-ethyl, ethyl urea, pellethane, athylisoharnstoff
PubChem CID 12254
IUPAC Name ethylurea
SMILES CCNC(N)=O

Specifications

Melting Point 88°C to 94°C
Quantity 500 g
Beilstein 1740512
Solubility Information Soluble in methanol (25mg/ml),and water (100mg/ml).
Formula Weight 88.11
Percent Purity 98%
Chemical Name or Material Ethylurea
Hazard Category H302+H312+H332-H335-H341
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
EINECSNumber 210-898-8
RTECSNumber YT2335000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only