Learn More
Ethyltriphenylphosphonium iodide, 98+%
CAS: 4736-60-1 | C20H20IP | 418.26 g/mol
Supplier: Thermo Scientific Chemicals A1753322
Description
Ethyltriphenylphosphonium iodide is involved in synthesis of diarylmethine derivatives, phosphonium salts, and bismuth(III) polynuclear halide complexes, asymmetric hydrogenation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyltriphenylphosphonium iodide is involved in synthesis of diarylmethine derivatives, phosphonium salts, and bismuth(III) polynuclear halide complexes, asymmetric hydrogenation.
Solubility
Soluble in water.
Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store away from oxidizing agents.
Specifications
| Ethyltriphenylphosphonium iodide | |
| 164°C to 169°C | |
| MFCD00040352 | |
| UN2811 | |
| Hygroscopic; Light sensitive | |
| Soluble in water. | |
| [I-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 418.26 | |
| 418.26 |
| 4736-60-1 | |
| C20H20IP | |
| 100 g | |
| 3659323 | |
| ethyltriphenylphosphonium iodide, phosphonium, ethyltriphenyl-, iodide, ethyl triphenylphosphonium iodide, phenylphosphonium ethyl iodide, unii-xxz2v239jn, xxz2v239jn, ethyltriphenylphosphanium iodide, phosphonium, ethyltriphenyl-, iodide 1:1, ethyl triphenyl phosphanium iodide, ethyl triphenyl phosphonium iodide | |
| SLAFUPJSGFVWPP-UHFFFAOYSA-M | |
| ethyl(triphenyl)phosphanium;iodide | |
| 78474 | |
| ≥98% |
Chemical Identifiers
| 4736-60-1 | |
| 418.26 | |
| SLAFUPJSGFVWPP-UHFFFAOYSA-M | |
| 78474 | |
| [I-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C20H20IP | |
| MFCD00040352 | |
| ethyltriphenylphosphonium iodide, phosphonium, ethyltriphenyl-, iodide, ethyl triphenylphosphonium iodide, phenylphosphonium ethyl iodide, unii-xxz2v239jn, xxz2v239jn, ethyltriphenylphosphanium iodide, phosphonium, ethyltriphenyl-, iodide 1:1, ethyl triphenyl phosphanium iodide, ethyl triphenyl phosphonium iodide | |
| ethyl(triphenyl)phosphanium;iodide |
Safety and Handling
GHS H Statement
H301-H318-H312-H335
Toxic if swallowed.
Causes serious eye damage.
Harmful in contact with skin.
May cause respiratory irritation.
P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H312-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 225-245-2
RTECSNumber : TA2312000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only