missing translation for 'onlineSavingsMsg'
Learn More

Ethyltriphenylphosphonium iodide, 98+%

Catalog No. AAA1753314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1753314 25 g
AAA1753322 100 g
AAA1753336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1753314 Supplier Thermo Scientific Chemicals Supplier No. A1753314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4736-60-1 | C20H20IP | 418.26 g/mol

Ethyltriphenylphosphonium iodide is involved in synthesis of diarylmethine derivatives, phosphonium salts, and bismuth(III) polynuclear halide complexes, asymmetric hydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyltriphenylphosphonium iodide is involved in synthesis of diarylmethine derivatives, phosphonium salts, and bismuth(III) polynuclear halide complexes, asymmetric hydrogenation.

Solubility
Soluble in water.

Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 4736-60-1
Molecular Formula C20H20IP
Molecular Weight (g/mol) 418.26
MDL Number MFCD00040352
InChI Key SLAFUPJSGFVWPP-UHFFFAOYSA-M
Synonym ethyltriphenylphosphonium iodide, phosphonium, ethyltriphenyl-, iodide, ethyl triphenylphosphonium iodide, phenylphosphonium ethyl iodide, unii-xxz2v239jn, xxz2v239jn, ethyltriphenylphosphanium iodide, phosphonium, ethyltriphenyl-, iodide 1:1, ethyl triphenyl phosphanium iodide, ethyl triphenyl phosphonium iodide
PubChem CID 78474
IUPAC Name ethyl(triphenyl)phosphanium;iodide
SMILES [I-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 164°C to 169°C
Quantity 25 g
UN Number UN2811
Beilstein 3659323
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in water.
Formula Weight 418.26
Percent Purity ≥98%
Chemical Name or Material Ethyltriphenylphosphonium iodide
Hazard Category H301-H312-H315-H319
Hazard Statement GHS H Statement
H301-H318-H312-H335
Toxic if swallowed.
Causes serious eye damage.
Harmful in contact with skin.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 225-245-2
RTECSNumber TA2312000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only