missing translation for 'onlineSavingsMsg'
Learn More

Ethylphosphonic dichloride, 98%, Thermo Scientific Chemicals

Catalog No. p-7023878
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1786906 5 g
AAA1786914 25 g
AAA1786922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1786906 Supplier Thermo Scientific Chemicals Supplier No. A1786906

Ethylphosphonic dichloride finds it uses in the preparation of: ethylphosphonic diisocyanate, 4-acetylphenyl isopropyl ethylphosphonate, 4-acetylphenyl phenyl ethylphosphonate and 4-acetylphenyl cyclohexyl ethylphosphonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylphosphonic dichloride finds it uses in the preparation of: ethylphosphonic diisocyanate, 4-acetylphenyl isopropyl ethylphosphonate, 4-acetylphenyl phenyl ethylphosphonate and 4-acetylphenyl cyclohexyl ethylphosphonate.

Solubility
Soluble in chloroform and dichloromethane.

Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1066-50-8
Molecular Formula C2H5Cl2OP
Molecular Weight (g/mol) 146.93
MDL Number MFCD00002073
InChI Key OWGJXSYVHQEVHS-UHFFFAOYSA-N
Synonym ethylphosphonic dichloride, phosphonic dichloride, ethyl, ethylphosphoryl dichloride, ethyldichlorophosphite, aurora ka-1351, acmc-20akvx, ethylphosphonyl dichloride, ethyl phosphonic dichloride, dichloroethylphosphine oxide, 1-dichloro-phosphoryl-ethane
PubChem CID 70596
IUPAC Name 1-dichlorophosphorylethane
SMILES CCP(Cl)(Cl)=O

Specifications

Density 1.376
Boiling Point 71°C to 72°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4665
Quantity 5 g
UN Number UN2922
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform and dichloromethane.
Formula Weight 146.94
Percent Purity 98%
Chemical Name or Material Ethylphosphonic dichloride
Show More Show Less
Hazard Category H302+H312+H332-H314
Hazard Statement GHS H Statement
H330-H314-H318
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 213-918-3
RTECSNumber TA1780000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only