accessibility menu, dialog, popup

UserName

H3CPClClO

CAS RN 1066-50-8

IUPAC Name: ethylphosphonoyl dichloride
Synonyms: phosphonic dichloride, ethyl-
Molecular Weight (g/mol): 146.93
Molecular Formula: C2H5Cl2OP
InChi Key: OWGJXSYVHQEVHS-UHFFFAOYSA-N
SMILES: CCP(Cl)(Cl)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)