missing translation for 'onlineSavingsMsg'
Learn More

Ethylparaben, NF, 98-102%, Spectrum™ Chemical

Catalog No. 18602580
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602580 2.5 kg Poly Bottle
18602578 125 g Poly Bottle
18602579 500 g Poly Bottle
18602581 12 kg Poly Bottle
18602582 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18602580 Supplier Spectrum Chemical Mfg Cor Supplier No. ET1172.5KG
Only null left

ET117, 120-47-8, C9H10O3

Spectrum™ Chemical Ethylparaben, NF is used as an antimicrobial preservative. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 120-47-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
InChI Key NUVBSKCKDOMJSU-UHFFFAOYSA-N
IUPAC Name ethyl 4-hydroxybenzoate
SMILES CCOC(=O)C1=CC=C(O)C=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Poly Bottle
Quantity 2.5 kg
Percent Purity 98 to 102%
Grade NF
Physical Form Crystals or Flakes