missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylmalonic Acid 98.0+%, TCI America™
$170.87 - $689.20
Chemical Identifiers
| CAS | 601-75-2 |
|---|---|
| Molecular Formula | C5H8O4 |
| Molecular Weight (g/mol) | 132.115 |
| MDL Number | MFCD00002668 |
| InChI Key | UKFXDFUAPNAMPJ-UHFFFAOYSA-N |
| Synonym | ethylmalonic acid, 2-ethylmalonic acid, propanedioic acid, ethyl, 1,1-propanedicarboxylic acid, alpha-carboxybutyric acid, malonic acid, ethyl, ethylmalonate, ethyl-malonic acid, unii-432nf49dfg, ethylpropanedioic acid |
| PubChem CID | 11756 |
| ChEBI | CHEBI:741548 |
| IUPAC Name | 2-ethylpropanedioic acid |
| SMILES | CCC(C(=O)O)C(=O)O |
Chemical Identifiers
| 601-75-2 | |
| 132.115 | |
| UKFXDFUAPNAMPJ-UHFFFAOYSA-N | |
| 11756 | |
| 2-ethylpropanedioic acid |
| C5H8O4 | |
| MFCD00002668 | |
| ethylmalonic acid, 2-ethylmalonic acid, propanedioic acid, ethyl, 1,1-propanedicarboxylic acid, alpha-carboxybutyric acid, malonic acid, ethyl, ethylmalonate, ethyl-malonic acid, unii-432nf49dfg, ethylpropanedioic acid | |
| CHEBI:741548 | |
| CCC(C(=O)O)C(=O)O |
Specifications
| 601-75-2 | |
| White | |
| MFCD00002668 | |
| ethylmalonic acid, 2-ethylmalonic acid, propanedioic acid, ethyl, 1,1-propanedicarboxylic acid, alpha-carboxybutyric acid, malonic acid, ethyl, ethylmalonate, ethyl-malonic acid, unii-432nf49dfg, ethylpropanedioic acid | |
| CCC(C(=O)O)C(=O)O | |
| 132.115 | |
| CHEBI:741548 | |
| ≥98.0% (T) | |
| Ethylmalonic Acid |
| 112°C | |
| C5H8O4 | |
| 5 g | |
| UKFXDFUAPNAMPJ-UHFFFAOYSA-N | |
| 2-ethylpropanedioic acid | |
| 11756 | |
| 132.12 | |
| Crystalline Powder |
Safety and Handling
TSCA : No