missing translation for 'onlineSavingsMsg'
Learn More

Ethylmalonic Acid 98.0+%, TCI America™

Supplier:  TCI America E013725G

 View more versions of this product

Catalog No. E013725G


Only null left

Description

Description

Specifications

Specifications

Ethylmalonic Acid
112°C
C5H8O4
25 g
UKFXDFUAPNAMPJ-UHFFFAOYSA-N
2-ethylpropanedioic acid
11756
132.12
Crystalline Powder
601-75-2
White
MFCD00002668
ethylmalonic acid, 2-ethylmalonic acid, propanedioic acid, ethyl, 1,1-propanedicarboxylic acid, alpha-carboxybutyric acid, malonic acid, ethyl, ethylmalonate, ethyl-malonic acid, unii-432nf49dfg, ethylpropanedioic acid
CCC(C(=O)O)C(=O)O
132.115
CHEBI:741548
≥98.0% (T)

Chemical Identifiers

601-75-2
132.115
UKFXDFUAPNAMPJ-UHFFFAOYSA-N
11756
2-ethylpropanedioic acid
C5H8O4
MFCD00002668
ethylmalonic acid, 2-ethylmalonic acid, propanedioic acid, ethyl, 1,1-propanedicarboxylic acid, alpha-carboxybutyric acid, malonic acid, ethyl, ethylmalonate, ethyl-malonic acid, unii-432nf49dfg, ethylpropanedioic acid
CHEBI:741548
CCC(C(=O)O)C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions