missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylhydrocupreine hydrochloride, MilliporeSigma™ Supelco™
Description
Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 3413-58-9 |
| Physical Form | Neat |
| Linear Formula | C21H28N2O2 · HCl |
| MDL Number | MFCD00135594 |
| Synonym | Optochin |
| Type | Analytical Standard |
| Quantity | 1 g |
| Formula Weight | 376.92 |
| Grade | Analytical Standard |