missing translation for 'onlineSavingsMsg'
Learn More

Ethylhydrocupreine hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111013969
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013969 1 g
111013971 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013969 Supplier Merck Emd Millipore Supplier No. E98761G
Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 3413-58-9
Physical Form Neat
Linear Formula C21H28N2O2 · HCl
MDL Number MFCD00135594
Synonym Optochin
Type Analytical Standard
Quantity 1 g
Formula Weight 376.92
Grade Analytical Standard