missing translation for 'onlineSavingsMsg'
Learn More

Ethylhexyl Triazone 98.0+%, TCI America™

$31.68 - $70.84

Chemical Identifiers

CAS 88122-99-0
Molecular Formula C48H66N6O6
Molecular Weight (g/mol) 823.092
MDL Number MFCD09753106
InChI Key JGUMTYWKIBJSTN-UHFFFAOYSA-N
Synonym Tris(2-ethylhexyl) 4,4′,4′′-(1,3,5-Triazine-2,4,6-triyltriimino)trisbenzoate, 4,4′,4′′-(1,3,5-Triazine-2,4,6-triyltriimino)trisbenzoic Acid Tris(2-ethylhexyl) Ester
PubChem CID 159201
IUPAC Name 2-ethylhexyl 4-[[4,6-bis[4-(2-ethylhexoxycarbonyl)anilino]-1,3,5-triazin-2-yl]amino]benzoate
SMILES CCCCC(CC)COC(=O)C1=CC=C(C=C1)NC2=NC(=NC(=N2)NC3=CC=C(C=C3)C(=O)OCC(CC)CCCC)NC4=CC=C(C=C4)C(=O)OCC(CC)CCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E13121G
View Documents
TCI America
E13121G
1 g
Each for $31.68
Only null left
 
E13125G
View Documents
TCI America
E13125G
5 g
Each for $70.84
Only null left
 
Description

Description

Specifications

Chemical Identifiers

88122-99-0
823.092
JGUMTYWKIBJSTN-UHFFFAOYSA-N
159201
CCCCC(CC)COC(=O)C1=CC=C(C=C1)NC2=NC(=NC(=N2)NC3=CC=C(C=C3)C(=O)OCC(CC)CCCC)NC4=CC=C(C=C4)C(=O)OCC(CC)CCCC
C48H66N6O6
MFCD09753106
Tris(2-ethylhexyl) 4,4′,4′′-(1,3,5-Triazine-2,4,6-triyltriimino)trisbenzoate, 4,4′,4′′-(1,3,5-Triazine-2,4,6-triyltriimino)trisbenzoic Acid Tris(2-ethylhexyl) Ester
2-ethylhexyl 4-[[4,6-bis[4-(2-ethylhexoxycarbonyl)anilino]-1,3,5-triazin-2-yl]amino]benzoate

Specifications

88122-99-0
White-Yellow
MFCD09753106
Tris(2-ethylhexyl) 4,4′,4′′-(1,3,5-Triazine-2,4,6-triyltriimino)trisbenzoate, 4,4′,4′′-(1,3,5-Triazine-2,4,6-triyltriimino)trisbenzoic Acid Tris(2-ethylhexyl) Ester
CCCCC(CC)COC(=O)C1=CC=C(C=C1)NC2=NC(=NC(=N2)NC3=CC=C(C=C3)C(=O)OCC(CC)CCCC)NC4=CC=C(C=C4)C(=O)OCC(CC)CCCC
823.092
823.09
Crystalline Powder
126°C
C48H66N6O6
1 g
JGUMTYWKIBJSTN-UHFFFAOYSA-N
2-ethylhexyl 4-[[4,6-bis[4-(2-ethylhexoxycarbonyl)anilino]-1,3,5-triazin-2-yl]amino]benzoate
159201
≥98.0% (N)
Ethylhexyl Triazone
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'