missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylhexyl Triazone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 126°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 823.09 |
| Percent Purity | ≥98.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Ethylhexyl Triazone |
Safety and Handling
| TSCA | No |