missing translation for 'onlineSavingsMsg'
Learn More

Ethylhexyl triazone, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015741
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015741 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015741 Supplier Merck Emd Millipore Supplier No. 93465100MG

Ethylhexyl triazone is a ultraviolet-B filter, which is used as a sunscreen agent in skin care products.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 88122-99-0
Flash Point 307°C
Physical Form Neat
Linear Formula C48H66N6O6
MDL Number MFCD09753106
Melting Point 128°C to 132°C
Synonym 2,4,6-Trianilino(p-carbo-2-ethylhexyl-1-oxy)-1,3,5-triazine; Tris(2-ethylhexyl) 4,4 ′,4 ′′-(1,3,5-triazine-2,4,6-triyltriimino)trisbenzoate
Type Analytical Standard
Quantity 100 mg
Formula Weight 823.07
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less