missing translation for 'onlineSavingsMsg'
Learn More

ethylethiomorpholine-3-carboxylate hydrochloride, 90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC27673DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC27673DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

159381-07-4
211.704
QQRCEZBURAQLBQ-UHFFFAOYSA-N
17433447
CCOC(=O)C1CSCCN1.Cl
C7H14ClNO2S
MFCD00107968
ethyl thiomorpholine-3-carboxylate hydrochloride, thiomorpholine-3-carboxylic acid ethyl ester hydrochloride, thiomorpholine-3-carboxylic acid ethyl ester hcl, thiomorpholine-3-carboxylicacidethylesterhydrochloride, ethyl thiomorpholine-3-carboxylate hcl, ethylthiomorpholine-3-carboxylatehydrochloride, 3-ethoxycarbonyl thiomorpholine hydrochloride, ethyl 1,4-thiazinane-3-carboxylate hydrochloride, ethyl 1,4-thiazaperhydroine-3-carboxylate, chloride, thiomorpholine-3-carboxylic acid ethyl esterhydrochloride
ethyl thiomorpholine-3-carboxylate;hydrochloride

Specifications

ethylethiomorpholine-3-carboxylate hydrochloride
89.5
90%
C7H14ClNO2S
MFCD00107968
QQRCEZBURAQLBQ-UHFFFAOYSA-N
ethyl thiomorpholine-3-carboxylate;hydrochloride
17433447
90%
159381-07-4
100.0
Amber Glass Bottle
1g
ethyl thiomorpholine-3-carboxylate hydrochloride, thiomorpholine-3-carboxylic acid ethyl ester hydrochloride, thiomorpholine-3-carboxylic acid ethyl ester hcl, thiomorpholine-3-carboxylicacidethylesterhydrochloride, ethyl thiomorpholine-3-carboxylate hcl, ethylthiomorpholine-3-carboxylatehydrochloride, 3-ethoxycarbonyl thiomorpholine hydrochloride, ethyl 1,4-thiazinane-3-carboxylate hydrochloride, ethyl 1,4-thiazaperhydroine-3-carboxylate, chloride, thiomorpholine-3-carboxylic acid ethyl esterhydrochloride
CCOC(=O)C1CSCCN1.Cl
211.704
211.71
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents