accessibility menu, dialog, popup

UserName

CAS RN 159381-07-4

H3COOSHNHCl

CAS RN 159381-07-4

IUPAC Name: ethyl thiomorpholine-3-carboxylate hydrochloride
Synonyms: ethyl thiomorpholine-3-carboxylate hydrochloride
Molecular Weight (g/mol): 211.7
Molecular Formula: C7H14ClNO2S
InChi Key: QQRCEZBURAQLBQ-UHFFFAOYNA-N
SMILES: Cl.CCOC(=O)C1CSCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)