missing translation for 'onlineSavingsMsg'
Learn More

Ethylenethiourea 98.0+%, TCI America™

Catalog No. I000425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I000425G 25 g
I0004500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I000425G Supplier TCI America Supplier No. I000425G

Chemical Identifiers

CAS 96-45-7
Molecular Formula C3H6N2S
Molecular Weight (g/mol) 102.16
MDL Number MFCD00005276
InChI Key PDQAZBWRQCGBEV-UHFFFAOYSA-N
Synonym 2-imidazolidinethione, ethylenethiourea, ethylene thiourea, 2-mercaptoimidazoline, imidazolidinethione, n,n'-ethylenethiourea, mercazin i, warecure c, rhenogran etu, mercaptoimidazoline
PubChem CID 2723650
ChEBI CHEBI:34750
IUPAC Name imidazolidine-2-thione
SMILES S=C1NCCN1

Specifications

Melting Point 198°C
Color White
Quantity 25 g
Formula Weight 102.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethylenethiourea
EINECSNumber (5)-0423
RTECSNumber NI9625000
TSCA Yes