missing translation for 'onlineSavingsMsg'
Learn More

Ethylenedithiobis(trimethylsilane) 97.0+%, TCI America™

Catalog No. E04795G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E04795G Supplier TCI America Supplier No. E04795G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51048-29-4
Molecular Formula C8H22S2Si2
Molecular Weight (g/mol) 238.554
MDL Number MFCD00042832
InChI Key CBRFJNLREFDKBD-UHFFFAOYSA-N
Synonym S,S′C-Bis(trimethylsilyl)-1,2-ethanedithiol, 1,2-Bis(trimethylsilylthio)ethane, 1,2-Ethanedithiobis(trimethylsilane)
PubChem CID 101207
IUPAC Name trimethyl(2-trimethylsilylsulfanylethylsulfanyl)silane
SMILES C[Si](C)(C)SCCS[Si](C)(C)C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 238.55
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylenedithiobis(trimethylsilane) [Protecting Reagent for Aldehydes and Ketones]