missing translation for 'onlineSavingsMsg'
Learn More

(Ethylenedithio)diacetic Acid 98.0+%, TCI America™

Catalog No. p-7128916
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E027825G 25 g
E0278100G 100 g
E0278500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E027825G Supplier TCI America Supplier No. E027825G
Only null left

Chemical Identifiers

CAS 7244-02-2
Molecular Formula C6H8O4S2
Molecular Weight (g/mol) 208.25
MDL Number MFCD00021771
InChI Key CCQMORVULNZXIA-UHFFFAOYSA-L
Synonym ethylenedithio diacetic acid, 1,2-bis carboxymethylthio ethane, 2-2-carboxymethylsulfanyl ethylsulfanyl acetic acid, 2-2-carboxymethylthio ethylthio acetic acid, 2-carboxymethyl sulfanyl ethyl sulfanyl acetic acid, ethylenedithiodiacetic acid, acmc-2097kv, ethylenebisthio diacetic acid, 1,2-ethylenebis thioglycolic acid, 2,2'-ethylenedithio diacetic acid
PubChem CID 310815
IUPAC Name 2-[2-(carboxymethylsulfanyl)ethylsulfanyl]acetic acid
SMILES C(CSCC(=O)O)SCC(=O)O

Specifications

Melting Point 110°C
Color White
Quantity 25 g
Formula Weight 210.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (Ethylenedithio)diacetic Acid