missing translation for 'onlineSavingsMsg'
Learn More

Ethylenedioxybis(trimethylsilane) 97.0+%, TCI America™

Catalog No. E047825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E047825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E047825G Supplier TCI America Supplier No. E047825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7381-30-8
Molecular Formula C8H22O2Si2
Molecular Weight (g/mol) 206.43
MDL Number MFCD00009640
InChI Key JGWFUSVYECJQDT-UHFFFAOYSA-N
Synonym 1,2-bis trimethylsiloxy ethane, 1,2-bis trimethylsilyloxy ethane, 2,2,7,7-tetramethyl-3,6-dioxa-2,7-disilaoctane, ethylene glycol bis trimethylsilyl ether, ethylenedioxybis trimethylsilane, 3,6-dioxa-2,7-disilaoctane, 2,2,7,7-tetramethyl, 1,2-bis-trimethylsilyloxy ethane, 1,2-bis-trimethylsilanyloxy-ethane, ethylene glycol, di-tms, acmc-1bh50
PubChem CID 81858
IUPAC Name 2,2,7,7-tetramethyl-3,6-dioxa-2,7-disilaoctane
SMILES C[Si](C)(C)OCCO[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 166°C
Quantity 25 g
UN Number 1993
Formula Weight 206.43
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylenedioxybis(trimethylsilane) [Protecting Reagent for Aldehydes and Ketones]