accessibility menu, dialog, popup

UserName

CAS RN 7381-30-8

IUPAC Name: 2,2,7,7-tetramethyl-3,6-dioxa-2,7-disilaoctane
Synonyms: 2,2,7,7-tetramethyl-3,6-dioxa-2,7-disilaoctane
Molecular Weight (g/mol): 206.43
Molecular Formula: C8H22O2Si2
InChi Key: JGWFUSVYECJQDT-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OCCO[Si](C)(C)C
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)