missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Dianhydride 98.0+%, TCI America™
Supplier: TCI America E048025G
Specifications
| Ethylenediaminetetraacetic Dianhydride | |
| 190°C | |
| MFCD00074963 | |
| 4,4′C-Ethylenebis(2,6-morpholinedione), EDTA Dianhydride | |
| C1C(=O)OC(=O)CN1CCN2CC(=O)OC(=O)C2 | |
| 256.214 | |
| 256.21 | |
| Crystalline Powder |
| 23911-25-3 | |
| C10H12N2O6 | |
| 25 g | |
| POLIXZIAIMAECK-UHFFFAOYSA-N | |
| 4-[2-(2,6-dioxomorpholin-4-yl)ethyl]morpholine-2,6-dione | |
| 513917 | |
| ≥98.0% (T) |
Chemical Identifiers
| 23911-25-3 | |
| 256.214 | |
| POLIXZIAIMAECK-UHFFFAOYSA-N | |
| 4-[2-(2,6-dioxomorpholin-4-yl)ethyl]morpholine-2,6-dione |
| C10H12N2O6 | |
| MFCD00074963 | |
| 513917 | |
| C1C(=O)OC(=O)CN1CCN2CC(=O)OC(=O)C2 |
Safety and Handling
EINECSNumber : (2)-1264
TSCA : No