accessibility menu, dialog, popup

UserName

OOONNOOO

CAS RN 23911-25-3

IUPAC Name: 4-[2-(2,6-dioxomorpholin-4-yl)ethyl]morpholine-2,6-dione
Synonyms: 4-[2-(2,6-dioxomorpholin-4-yl)ethyl]morpholine-2,6-dione
Molecular Weight (g/mol): 256.21
Molecular Formula: C10H12N2O6
InChi Key: POLIXZIAIMAECK-UHFFFAOYSA-N
SMILES: O=C1CN(CCN2CC(=O)OC(=O)C2)CC(=O)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results