missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Tetrasodium Salt, Solution, Spectrum™ Chemical

Catalog No. p-7157375
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Drum
Poly Bottle
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602574 1 L Poly Bottle
18602575 4 L Poly Bottle
18602576 20 L Poly Drum
18602577 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602574 Supplier Spectrum Chemical Mfg Cor Supplier No. E10481LT
Only null left

E1048, 13235-36-4, C10H12N2O8Na4

Spectrum™ Chemical Ethylenediaminetetraacetic Acid Tetrasodium Salt, Solution is typically known as EDTA and is an aminopolycarboxylic acid. It is usually used to dissolve limescale. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 64-02-8 , 7732-18-5
Molecular Formula C10H12N2Na4O8
Molecular Weight (g/mol) 380.17
InChI Key UEUXEKPTXMALOB-UHFFFAOYSA-J
IUPAC Name tetrasodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
SMILES [Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

CAS Max % 38%, 56%
Packaging Poly Bottle
Quantity 1 L
UN Number UN3267
Grade Reagent