missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid Tetrasodium Salt, Dihydrate, Reagent, 98%, Spectrum™ Chemical
E1047, 10378-23-1, C10H12N2O8Na4·2H2O
Supplier: Spectrum Chemical Mfg Cor E104712KG
Description
Spectrum™ Chemical Ethylenediaminetetraacetic Acid Tetrasodium Salt, Dihydrate, Reagent is a solid that has no color and is water soluble. It is mainly used to dissolve lime scale but has many other uses in fields such as medicine, industry, cosmetics and laboratories as a chelating agent and sequestrant. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.Specifications
| 10378-23-1 | |
| Poly Pail | |
| 12 kg | |
| O.O.[Na].[Na].[Na].[Na].OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O | |
| 420.23 | |
| Reagent |
| 100% | |
| C10H20N2Na4O10 | |
| DGWOWDJDIBPYOR-UHFFFAOYSA-N | |
| 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid dihydrate tetrasodium | |
| 98% |
Chemical Identifiers
| 10378-23-1 | |
| 420.23 | |
| 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid dihydrate tetrasodium |
| C10H20N2Na4O10 | |
| DGWOWDJDIBPYOR-UHFFFAOYSA-N | |
| O.O.[Na].[Na].[Na].[Na].OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |