missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Tetrasodium Salt, Dihydrate, Reagent, 98%, Spectrum™ Chemical

Catalog No. 18602570
Change view
Click to view available options
Quantity:
125 g
500 g
3 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602570 500 g Poly Bottle
18602569 125 g Poly Bottle
18602571 3 kg Poly Pail
18602572 12 kg Poly Pail
18602573 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18602570 Supplier Spectrum Chemical Mfg Cor Supplier No. E1047500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

E1047, 10378-23-1, C10H12N2O8Na4·2H2O

Spectrum™ Chemical Ethylenediaminetetraacetic Acid Tetrasodium Salt, Dihydrate, Reagent is a solid that has no color and is water soluble. It is mainly used to dissolve lime scale but has many other uses in fields such as medicine, industry, cosmetics and laboratories as a chelating agent and sequestrant. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 10378-23-1
Molecular Formula C10H20N2Na4O10
Molecular Weight (g/mol) 420.23
InChI Key DGWOWDJDIBPYOR-UHFFFAOYSA-N
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid dihydrate tetrasodium
SMILES O.O.[Na].[Na].[Na].[Na].OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 500 g
Percent Purity 98%
Grade Reagent