missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%

Catalog No. p-4813481
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446081000 100 g Plastic bottle
AC446085000 500 g Plastic bottle
AC446080010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC446081000 Supplier Thermo Scientific Chemicals Supplier No. 446081000
Only null left

CAS: 10378-23-1 | C10H12N2Na4O8·2H2O | 416.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent

Chemical Identifiers

CAS 10378-23-1
Molecular Formula C10H12N2Na4O8·2H2O
Molecular Weight (g/mol) 416.21
InChI Key DGWOWDJDIBPYOR-UHFFFAOYSA-N
Synonym ethylenediaminetetraacetic acid tetrasodium salt dihydrate, tetrasodium ethylenediaminetetraacetate dihydrate, edta-na4, edta tetrasodium salt, edta tetrasodium salt hydrate, ethylenediaminetetraacetic acid tetrasodium salt hydrate, 64-02-8 anhydrous, ksc909a7h, edta tetrasodium salt dihydrate, tetrapotassium ion 4-edta dihydrate
PubChem CID 131843362
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;dihydrate
SMILES C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.O.[Na].[Na].[Na].[Na]

Specifications

Melting Point 300.0°C
Assay Percent Range 99.0 to 102% (on dry substance) (Complexometry)
Packaging Plastic bottle
Quantity 100 g
Solubility Information (16% in water) Clear solution
Formula Weight 416.21
Percent Purity 99%
Chemical Name or Material Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only