missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid 98.0+%, TCI America™
Supplier: TCI America E0084500G
Specifications
| Ethylenediaminetetraacetic Acid | |
| 240°C | |
| C10H16N2O8 | |
| 500 g | |
| KCXVZYZYPLLWCC-UHFFFAOYSA-N | |
| 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid | |
| 6049 | |
| 292.24 | |
| Crystalline Powder |
| 60-00-4 | |
| White | |
| MFCD00003541 | |
| (Ethylenedinitrilo)tetraacetic Acid, EDTA | |
| OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O | |
| 292.24 | |
| CHEBI:42191 | |
| ≥98.0% (T) |
Chemical Identifiers
| 60-00-4 | |
| 292.24 | |
| KCXVZYZYPLLWCC-UHFFFAOYSA-N | |
| 6049 | |
| 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid |
| C10H16N2O8 | |
| MFCD00003541 | |
| (Ethylenedinitrilo)tetraacetic Acid, EDTA | |
| CHEBI:42191 | |
| OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |
Safety and Handling
EINECSNumber : (2)-1263&(2)-1296
RTECSNumber : AH4025000
TSCA : Yes