missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid 98.0+%, TCI America™

Catalog No. E008425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E008425G 25 g
E0084500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E008425G Supplier TCI America Supplier No. E008425G
Only null left

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym (Ethylenedinitrilo)tetraacetic Acid, EDTA
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Melting Point 240°C
Color White
Quantity 25 g
Formula Weight 292.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic Acid
EINECSNumber (2)-1263&(2)-1296
RTECSNumber AH4025000
TSCA Yes