missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Monosodium Ferric Salt Hydrate 90.0+%, TCI America™

Catalog No. E009225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E009225G 25 g
E0092500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E009225G Supplier TCI America Supplier No. E009225G

Chemical Identifiers

CAS 15708-41-5
Molecular Formula C10H12FeN2NaO8
Molecular Weight (g/mol) 367.047
MDL Number MFCD00078215
InChI Key MKWYFZFMAMBPQK-UHFFFAOYSA-J
Synonym sodium feredetate, edta ferric sodium salt, calmosine, sodium ironedetate, edta iron iii sodium salt, ferisan, sytron, sequestrene nafe, sodium iron edta, ferric sodium edta
PubChem CID 27461
ChEBI CHEBI:78292
IUPAC Name sodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;iron(3+)
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Na+].[Fe+3]

Specifications

Quantity 25 g
Formula Weight 367.05
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic Acid Monosodium Ferric Salt Hydrate
EINECSNumber (2)-1266
RTECSNumber AH4900000
TSCA Yes