missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid manganese disodium salt hydrate

Catalog No. p-7051128
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AA4051906 5 g
AA4051914 25 g
2 options

Catalog No. AA4051906

Supplier: Thermo Scientific Chemicals 04051906

Only null left

Metal chelating compound | CAS: 15375-84-5 | C10H12MnN2Na2O8 | 389.13 g/mol

Ethylenediaminetetraacetic acid manganese disodium salt hydrate is a metal chelating compound. Cobalt was analysed complexometrically using ethylenediaminetetra-acetic acid (EDTA) by masking aluminum. Previous studies of manganese-EDTA and manganese-EGT-1 complexes indicated that the &-nitrogen coordination of EDTA resulted in a highly anisotropic spectrum for Mn(II). Lipophilic derivatives of manganese EDTA chelates when incorporated into liposomes confer the greatest hepatic enhancement per micromole of metal ion and have favorable clearance kinetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15375-84-5
Molecular Formula C10H12MnN2Na2O8
Molecular Weight (g/mol) 389.13
MDL Number MFCD00661156
InChI Key ZQHKAIDDHTYINE-UHFFFAOYSA-J
Synonym disodium manganese edta, edta disodium manganese salt, disodium manganese ethylenediaminetetraacetate, manganese disodium ethylene diamine tetraacetate, ethylenediaminetetraacetic acid, disodium manganese salt, disodium ethylenedinitrilo tetraacetato manganese, manganate 2-, disodium, glycine,n'-1,2-ethanediylbis n-carboxymethyl-, manganese disodium salt, manganate 2-,n'-1,2-ethanediylbis n-carboxymethyl glycinato 4--n,n',o,o',on,on'-, disodium, oc-6-21
PubChem CID 5148022
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;manganese(2+)
SMILES [Na+].[Na+].[Mn++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

Odor Odorless
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 389.13 (Anhydrous)
Physical Form Powder
Chemical Name or Material Ethylenediaminetetraacetic acid manganese disodium salt hydrate
EINECSNumber 239-407-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only