accessibility menu, dialog, popup

UserName

CAS RN 15375-84-5

NNOOOOOOOOMnNaNa

CAS RN 15375-84-5

IUPAC Name: manganese(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Synonyms: edta, disodium manganese salt|cyclohexanediamine tetraacetic acid
Molecular Weight (g/mol): 389.13
Molecular Formula: C10H12MnN2Na2O8
InChi Key: ZQHKAIDDHTYINE-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Mn++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results