missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Lead(II) Disodium Salt Hydrate 98.0+%, TCI America™

Catalog No. E009325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E009325G 25 g
E0093500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E009325G Supplier TCI America Supplier No. E009325G

Chemical Identifiers

CAS 22904-40-1
Molecular Formula C10H12N2Na2O8Pb
Molecular Weight (g/mol) 541.40
MDL Number MFCD00661155
InChI Key BIYQFSIBOJWRMG-UHFFFAOYSA-J
Synonym Lead(II) Disodium Ethylenediaminetetraacetate
PubChem CID 131876306
IUPAC Name disodium 2-[9-(carboxylatomethyl)-4,11-dioxo-1,3-dioxa-6,9-diaza-2-plumbacycloundecan-6-yl]acetate
SMILES [Na+].[Na+].[O-]C(=O)CN1CCN(CC([O-])=O)CC(=O)O[Pb]OC(=O)C1

Specifications

Color White
Quantity 25 g
UN Number 2291
Formula Weight 541.39
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic Acid Lead(II) Disodium Salt Hydrate
RTECSNumber AH4320000
TSCA Yes