CAS RN 22904-40-1
IUPAC Name:
disodium 2-[9-(carboxylatomethyl)-4,11-dioxo-1,3-dioxa-6,9-diaza-2-plumbacycloundecan-6-yl]acetate
Synonyms:
disodium 2-[9-(carboxylatomethyl)-4,11-dioxo-1,3-dioxa-6,9-diaza-2-plumbacycloundecan-6-yl]acetate
Molecular Weight (g/mol):
541.4
Molecular Formula:
C10H12N2Na2O8Pb
InChi Key:
BIYQFSIBOJWRMG-UHFFFAOYSA-J
SMILES:
[Na+].[Na+].[O-]C(=O)CN1CCN(CC([O-])=O)CC(=O)O[Pb]OC(=O)C1
1
–
1
of
1
results
1
Ethylenediaminetetraacetic Acid Lead(II) Disodium Salt Hydrate 98.0+%, TCI America™
CAS: 22904-40-1 Molecular Formula: C10H12N2Na2O8Pb Molecular Weight (g/mol): 541.40 MDL Number: MFCD00661155 InChI Key: BIYQFSIBOJWRMG-UHFFFAOYSA-J Synonym: Lead(II) Disodium Ethylenediaminetetraacetate PubChem CID: 131876306 IUPAC Name: disodium 2-[9-(carboxylatomethyl)-4,11-dioxo-1,3-dioxa-6,9-diaza-2-plumbacycloundecan-6-yl]acetate SMILES: [Na+].[Na+].[O-]C(=O)CN1CCN(CC([O-])=O)CC(=O)O[Pb]OC(=O)C1