missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid, iron(III) monosodium salt

Catalog No. p-7080446
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8899522 100 g
AA8899536 500 g
AA88995A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA8899522 Supplier Thermo Scientific Chemicals Supplier No. 08899522
Only null left

CAS: 15708-41-5 | C10H12FeN2NaO8 | 367.047 g/mol

Ethylenediaminetetraacetic acid, iron(III) monosodium salt is used to eliminate enzyme inhibition by traces of heavy metals and to inhibit enzymes that require divalent cations as cofactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15708-41-5
Molecular Formula C10H12FeN2NaO8
Molecular Weight (g/mol) 367.047
MDL Number MFCD00078215
InChI Key MKWYFZFMAMBPQK-UHFFFAOYSA-J
Synonym sodium feredetate, edta ferric sodium salt, calmosine, sodium ironedetate, edta iron iii sodium salt, ferisan, sytron, sequestrene nafe, sodium iron edta, ferric sodium edta
PubChem CID 27461
ChEBI CHEBI:78292
IUPAC Name sodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;iron(3+)
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Na+].[Fe+3]

Specifications

Odor Odorless
Quantity 100 g
Solubility Information Soluble in water.
Formula Weight 367.05
Physical Form Powder
Chemical Name or Material Ethylenediaminetetraacetic acid, iron (III) monosodium salt
EINECSNumber 239-802-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only