missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid, Honeywell Fluka™

Catalog No. p-7071708
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001815 500 g
6001814 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001815 Supplier Honeywell Chemicals Supplier No. 03610500G
Only null left

for complexometry,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Ignition Residue ≤0.1% (as SO4)
Packaging Plastic Bottle
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2H)2
Quantity 500 g
Beilstein 1716295
Solubility Information Soluble in water (0.4g/L at 20°C)
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
Formula Weight 292.24g/mol
Percent Purity ≥99%
Chemical Name or Material Ethylenediaminetetraacetic Acid
Hazard Statement P305 + P351 + P338
Precautionary Statement H319
EINECSNumber 200-449-4
RTECSNumber AH4025000