missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Ignition Residue | ≤0.1% (as SO4) |
| Packaging | Plastic Bottle |
| Linear Formula | (HO2CCH2)2NCH2CH2N(CH2CO2H)2 |
| Quantity | 1 kg |
| Beilstein | 1716295 |
| Solubility Information | Soluble in water (0.4g/L at 20°C) |
| IUPAC Name | 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid |
| Formula Weight | 292.24g/mol |
| Percent Purity | ≥99% |
| Chemical Name or Material | Ethylenediaminetetraacetic Acid |
Safety and Handling
| Hazard Statement | P305 + P351 + P338 |
| Precautionary Statement | H319 |
| EINECSNumber | 200-449-4 |
| RTECSNumber | AH4025000 |