missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid, Electrophoresis Grade, 500g, MP Biomedicals

Catalog No. ICN800682
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN800682 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN800682 Supplier MP Biomedicals Inc Supplier No. 04800682
Only null left

60-00-4 (EDTA) Purity: ≥99.5% Heavy metals (as Pb): <<10 ppm Iron (Fe): <<10 ppm Sulfated ash: <<0.1%

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Quantity 500 g
Chemical Name or Material Ethylenediaminetetraacetic Acid