missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid Disodium Salt Solution, 0.2M, Honeywell Fluka™
Volumetric, 0.2Â M EDTA-Na2, for complexometry
$309.63 - $309.63
Specifications
| Chemical Name or Material | Ethylenediaminetetraacetic acid disodium salt solution |
|---|---|
| Molecular Formula | C10H14N2Na2O8 |
| MDL Number | MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 |
| UN Number | NONH for all modes of transport |
| Beilstein | 3822669 |
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
| Ethylenediaminetetraacetic acid disodium salt solution | |
| MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 | |
| 3822669 | |
| ZGTMUACCHSMWAC-UHFFFAOYSA-L | |
| 336.21 | |
| CHEBI:64734 |
| C10H14N2Na2O8 | |
| NONH for all modes of transport | |
| ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid | |
| [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
| 57339238 |