missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid disodium salt dihydrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015017
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015017 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015017 Supplier Merck Emd Millipore Supplier No. PHR10681G
Only null left

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Pharmaceutical (small molecule)

Specifications

CAS 6381-92-6
Linear Formula C10H14N2 Na2O8 · 2H2 O
Melting Point 248°C
Synonym Edetate Disodium; Edetate disodium dihydrate; Disodium ethylenediaminetetraacetate dihydrate; EDTA disodium salt; EDTA-Na2; Edathamil; Edetate disodium salt dihydrate; Sequestrene Na2
Type Certified Reference Material
Quantity 1 g
Formula Weight 372.24
Grade Certified Reference Material