missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid disodium salt, 0.115N (0.0575M) Standardized solution

Catalog No. AA39782K2
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39782K2 1 L
AA39782K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA39782K2 Supplier Thermo Scientific Chemicals Supplier No. 039782K2

CAS: 139-33-3 | C10H14N2Na2O8 | 336.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 139-33-3
Molecular Formula C10H14N2Na2O8
Molecular Weight (g/mol) 336.21
MDL Number MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
InChI Key ZGTMUACCHSMWAC-UHFFFAOYSA-L
Synonym ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
PubChem CID 57339238
ChEBI CHEBI:64734
IUPAC Name disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
SMILES [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Concentration 0.115N (0.0575M)
Formula Weight 336.22
Odor Odorless
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Ethylenediaminetetraacetic acid disodium salt, Standardized Solution
EINECSNumber 205-358-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only