missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid Disodium Nickel(II) Salt Hydrate, TCI America™
Supplier: TCI America E009725G
Specifications
| Ethylenediaminetetraacetic Acid Disodium Nickel(II) Salt Hydrate | |
| Blue | |
| MFCD00070493 | |
| Nickel(II) Disodium Ethylenediaminetetraacetate | |
| C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.[Na+].[Na+].[Ni] | |
| 394.901 | |
| 392.88 |
| 15708-55-1 | |
| C10H14N2Na2NiO8 | |
| 25 g | |
| HRYMWOFNKDBTSQ-UHFFFAOYSA-L | |
| disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;nickel | |
| 102601958 | |
| Crystalline Powder |
Chemical Identifiers
| 15708-55-1 | |
| 394.901 | |
| HRYMWOFNKDBTSQ-UHFFFAOYSA-L | |
| 102601958 | |
| C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.[Na+].[Na+].[Ni] |
| C10H14N2Na2NiO8 | |
| MFCD00070493 | |
| Nickel(II) Disodium Ethylenediaminetetraacetate | |
| disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;nickel |
Safety and Handling
EINECSNumber : (2)-1266
RTECSNumber : QR6126600
TSCA : No