accessibility menu, dialog, popup

UserName

CAS RN 15708-55-1

NOOOONOOHOOHNaNaNi

CAS RN 15708-55-1

IUPAC Name: disodium 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxylatomethyl)amino)acetate nickel
Synonyms: disodium 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxylatomethyl)amino)acetate nickel
Molecular Weight (g/mol): 394.9
Molecular Formula: C10H14N2Na2NiO8
InChi Key: HRYMWOFNKDBTSQ-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[Ni].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results