Learn More
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate
CAS: 194491-32-2 | C10H12MgN2Na2O8 | 358.50 g/mol
$58.78 - $151.82
Chemical Identifiers
| CAS | 194491-32-2 |
|---|---|
| Molecular Formula | C10H12MgN2Na2O8 |
| Molecular Weight (g/mol) | 358.50 |
| MDL Number | MFCD00078217 |
| InChI Key | AWNVVAMWLMUZOZ-UHFFFAOYSA-J |
| Synonym | unii-ndt563s5vz, edta magnesium disodium, magnesium disodium edta, ndt563s5vz, magnesium disodium ethylenediaminetetraacetate, edta magnesium disodium salt hydrate, ethylenediaminetetraacetic acid magnesium disodium salt, ethylenediaminetetraacetic acid disodium magnesium salt, ethylenediaminetetraacetic acid disodium magnesium salt hydrate, magnesium sodium ethylenediaminetetraacetate |
| PubChem CID | 161064 |
| IUPAC Name | magnesium;disodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate |
| SMILES | [Na+].[Na+].[Mg++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1185714
|
Thermo Scientific Chemicals
L1185714 |
25 g |
Each for $58.78
|
|
|||||
|
AAL1185722
|
Thermo Scientific Chemicals
L1185722 |
100 g |
Each for $151.82
|
|
|||||
Description
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is used as chelating agents over a broad pH range in aqueous systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthylenediaminetetraacetic acid disodium magnesium salt hydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is used as chelating agents over a broad pH range in aqueous systems.
Solubility
Soluble in water. Insoluble in acid and organic liquids
Notes
Incompatible with oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Store at room temperature.
Chemical Identifiers
| 194491-32-2 | |
| 358.50 | |
| AWNVVAMWLMUZOZ-UHFFFAOYSA-J | |
| 161064 | |
| [Na+].[Na+].[Mg++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O |
| C10H12MgN2Na2O8 | |
| MFCD00078217 | |
| unii-ndt563s5vz, edta magnesium disodium, magnesium disodium edta, ndt563s5vz, magnesium disodium ethylenediaminetetraacetate, edta magnesium disodium salt hydrate, ethylenediaminetetraacetic acid magnesium disodium salt, ethylenediaminetetraacetic acid disodium magnesium salt, ethylenediaminetetraacetic acid disodium magnesium salt hydrate, magnesium sodium ethylenediaminetetraacetate | |
| magnesium;disodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate |
Specifications
| 194491-32-2 | |
| Odorless | |
| MFCD00078217 | |
| unii-ndt563s5vz, edta magnesium disodium, magnesium disodium edta, ndt563s5vz, magnesium disodium ethylenediaminetetraacetate, edta magnesium disodium salt hydrate, ethylenediaminetetraacetic acid magnesium disodium salt, ethylenediaminetetraacetic acid disodium magnesium salt, ethylenediaminetetraacetic acid disodium magnesium salt hydrate, magnesium sodium ethylenediaminetetraacetate | |
| AWNVVAMWLMUZOZ-UHFFFAOYSA-J | |
| magnesium;disodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate | |
| 161064 | |
| Ethylenediaminetetraacetic acid disodium magnesium salt hydrate |
| >300°C | |
| C10H12MgN2Na2O8 | |
| 25 g | |
| Soluble in water. Insoluble in acid and organic liquids | |
| [Na+].[Na+].[Mg++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O | |
| 358.50 | |
| 358.50 (Anhydrous) |
Safety and Handling
EINECSNumber : 238-372-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only