missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic acid diammonium salt hydrate
CAS: 20824-56-0 | C10H24N4O9 | 344.321 g/mol
$64.96 - $529.59
Chemical Identifiers
| CAS | 20824-56-0 |
|---|---|
| Molecular Formula | C10H24N4O9 |
| Molecular Weight (g/mol) | 344.321 |
| MDL Number | MFCD00150463 |
| InChI Key | WRSUMHYBOYWMTD-UHFFFAOYSA-N |
| Synonym | ethylenediaminetetraacetic acid diammonium salt hydrate, edetic acid diamine hydrate, diazanium,2-2-bis carboxymethyl amino ethyl-carboxylatome, ammonium 2,2-2-bis carboxymethyl amino ethyl azanediyl diacetate hydrate |
| PubChem CID | 71311375 |
| IUPAC Name | azane;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;hydrate |
| SMILES | C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.N.N.O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1528614
|
Thermo Scientific Chemicals
A1528614 |
25 g |
Each for $64.96
|
|
|||||
|
AAA1528622
|
Thermo Scientific Chemicals
A1528622 |
100 g |
Each for $149.70
|
|
|||||
|
AAA1528636
|
Thermo Scientific Chemicals
A1528636 |
500 g |
Each for $529.59
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 20824-56-0 | |
| 344.321 | |
| WRSUMHYBOYWMTD-UHFFFAOYSA-N | |
| 71311375 | |
| C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.N.N.O |
| C10H24N4O9 | |
| MFCD00150463 | |
| ethylenediaminetetraacetic acid diammonium salt hydrate, edetic acid diamine hydrate, diazanium,2-2-bis carboxymethyl amino ethyl-carboxylatome, ammonium 2,2-2-bis carboxymethyl amino ethyl azanediyl diacetate hydrate | |
| azane;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;hydrate |
Specifications
| 20824-56-0 | |
| MFCD00150463 | |
| 7785165 | |
| WRSUMHYBOYWMTD-UHFFFAOYSA-N | |
| azane;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;hydrate | |
| 71311375 | |
| Ethylenediaminetetraacetic acid diammonium salt hydrate |
| C10H24N4O9 | |
| 25 g | |
| ethylenediaminetetraacetic acid diammonium salt hydrate, edetic acid diamine hydrate, diazanium,2-2-bis carboxymethyl amino ethyl-carboxylatome, ammonium 2,2-2-bis carboxymethyl amino ethyl azanediyl diacetate hydrate | |
| C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.N.N.O | |
| 344.321 | |
| 326.31 (Anhydrous) |
Safety and Handling
EINECSNumber : 244-063-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only