missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, Cell Culture Reagent

Catalog No. AAJ6294836
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6294836 500 g
AAJ62948A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6294836 Supplier Thermo Scientific Chemicals Supplier No. J6294836

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol

Ethylenediaminetetraacetic acid solution can be used to study cell biology, molecular biology, bioactive small molecules, biochemicals, solutions and reagents, inorganic salts, chelators and ethylenediaminetetraacetic acid (EDTA) and EDTA solutions. Ethylenediaminetetraacetic acid solution has been used in a study to help elucidate the role of antigen retrieval in immunostaining of ethanol-fixed smears. Ethylenediaminetetraacetic acid solution has also been used in a study to assess the uptake and release of biotin-labeled fluorescein (b-FITC) as well as immobilization of biotin-labeled glucose oxidase (b-GOx) to the polyelectrolyte microcapsules with an avidin-poly(styrene sulfonate) (PSS) membrane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediaminetetraacetic acid solution can be used to study cell biology, molecular biology, bioactive small molecules, biochemicals, solutions and reagents, inorganic salts, chelators and ethylenediaminetetraacetic acid (EDTA) and EDTA solutions. Ethylenediaminetetraacetic acid solution has been used in a study to help elucidate the role of antigen retrieval in immunostaining of ethanol-fixed smears. Ethylenediaminetetraacetic acid solution has also been used in a study to assess the uptake and release of biotin-labeled fluorescein (b-FITC) as well as immobilization of biotin-labeled glucose oxidase (b-GOx) to the polyelectrolyte microcapsules with an avidin-poly(styrene sulfonate) (PSS) membrane.

Solubility
Soluble in water.

Notes
Not tested for endtoxin or mycoplasma contaminants

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Melting Point ∽245°C (decomposition)
Color White
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2H)2
Quantity 500 g
Beilstein 1716295
Merck Index 14,3517
Solubility Information Soluble in water.
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
Formula Weight 292.24
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material Ethylenediaminetetraacetic acid
Show More Show Less
Hazard Category H319-H332-H373
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P260-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P314-P501c
EINECSNumber 200-449-4
RTECSNumber AH4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only