missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid, 99%

Catalog No. p-7080950
Change view
Click to view available options
Quantity:
500 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1071336 500 g
AAA107130B 1000 g
AAA107130I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1071336 Supplier Thermo Scientific Chemicals Supplier No. A1071336
Only null left

CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol

Ethylenediaminetetraacetic acid is used as a complexing agent for many cations. It prevents metal ion impurities and used to remove excess iron from the body. It acts as a sequestering agent in cosmetic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediaminetetraacetic acid is used as a complexing agent for many cations. It prevents metal ion impurities and used to remove excess iron from the body. It acts as a sequestering agent in cosmetic products.

Solubility
Soluble in sodium hydroxide, sodium carbonate, ammonia solution, acid and common organic solvents. Slightly soluble in water.

Notes
Stable. Incompatible with copper, copper alloys, nickel, aluminum, strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Melting Point ∼245°C (decomposition)
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2H)2
Quantity 500 g
Beilstein 1716295
Merck Index 14,3517
Solubility Information Soluble in sodium hydroxide,sodium carbonate,ammonia solution,acid and common organic solvents. Slightly soluble in water.
Formula Weight 292.24
Percent Purity 99%
Chemical Name or Material Ethylenediaminetetraacetic acid
Hazard Category H319-H332-H373
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P260-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P314-P501c
EINECSNumber 200-449-4
RTECSNumber AH4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only