missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, 99.5%, ACS reagent

Catalog No. p-129284
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC409930010 1 kg Plastic bottle
AC409930050 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC409930010 Supplier Thermo Scientific Chemicals Supplier No. 409930010
Only null left

CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Decomposition Information >150°C
Melting Point 220.0°C
Color White
Flash Point >100°C
Ignition Residue 0.2% max.
Infrared Spectrum Authentic
Assay Percent Range 99.50%
Packaging Plastic bottle
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2H)2
Quantity 1 kg
Fieser 01,373
Merck Index 15, 3565
Solubility Information 0.005% max. Insoluble Matter (in dilute NH4OH)
Vapor Pressure 0.013hPa at 20°C
Formula Weight 292.23
Percent Purity 99.5%
Grade ACS Reagent
Physical Form Fine Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic acid
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-449-4
RTECSNumber AH4025000
TSCA TSCA

RUO – Research Use Only