missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic Acid, 0.5M aq. soln., pH 8.0, autoclaved

Catalog No. p-7041794
Change view
Click to view available options
Quantity:
250 mL
500 mL
Catalog No. Quantity
AAJ60292AK 250 mL
AAJ60292AP 500 mL
2 options

Catalog No. AAJ60292AK

Supplier: Thermo Scientific Chemicals J60292AK

Only null left

CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol

Ethylenediaminetetraacetic acid is used in the treatment of lead poisoning, brain damage, cancer, rheumatoid arthritis, osteoarthritis, an eye condition called macular degeneration, diabetes, Alzheimer's disease, multiple sclerosis and Parkinson's disease. It acts as a chelating agent and used for the preparation of many solutions including DNA loading dye and resuspension buffer which is isolation of plasmid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases, metals and copper.

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

pH 8.0
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2H)2
Quantity 250 mL
Solubility Information Miscible with water.
Formula Weight 292.24
Concentration or Composition (by Analyte or Components) 0.5M aq. soln.
Physical Form Liquid
Chemical Name or Material Ethylenediaminetetraacetic acid, autoclaved
Hazard Category H319-H373
Precautionary Statement P260-P264b-P280i-P305+P351+P338-P314-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only