missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, 0.5M aq. soln, pH 8.0

Catalog No. AAJ62786AK
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62786AK 250 mL
AAJ62786AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62786AK Supplier Thermo Scientific Chemicals Supplier No. J62786AK

CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol

Ethylenediaminetetraacetic acid solution can be used to study cell biology, molecular biology, bioactive small molecules, biochemicals, solutions and reagents, inorganic salts, chelators and ethylenediaminetetraacetic acid (EDTA) and EDTA solutions. Ethylenediaminetetraacetic acid solution has been used in a study to help elucidate the role of antigen retrieval in immunostaining of ethanol-fixed smears. Ethylenediaminetetraacetic acid solution has also been used in a study to assess the uptake and release of biotin-labeled fluorescein (b-FITC) as well as immobilization of biotin-labeled glucose oxidase (b-GOx) to the polyelectrolyte microcapsules with an avidin-poly(styrene sulfonate) (PSS) membrane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediaminetetraacetic acid solution can be used to study cell biology, molecular biology, bioactive small molecules, biochemicals, solutions and reagents, inorganic salts, chelators and ethylenediaminetetraacetic acid (EDTA) and EDTA solutions. Ethylenediaminetetraacetic acid solution has been used in a study to help elucidate the role of antigen retrieval in immunostaining of ethanol-fixed smears. Ethylenediaminetetraacetic acid solution has also been used in a study to assess the uptake and release of biotin-labeled fluorescein (b-FITC) as well as immobilization of biotin-labeled glucose oxidase (b-GOx) to the polyelectrolyte microcapsules with an avidin-poly(styrene sulfonate) (PSS) membrane.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Color Colorless
pH 8.0
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2H)2
Quantity 250 mL
Solubility Information Soluble in water.
Formula Weight 292.24
Concentration or Composition (by Analyte or Components) 0.5M aq. soln.
Physical Form Liquid
Chemical Name or Material Ethylenediaminetetraacetic acid
Hazard Category H319-H373
Precautionary Statement P260-P264b-P280i-P305+P351+P338-P314-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only