missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine tetraacetic acid disodium salt dihydrate, 99.74%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219517391

 View more versions of this product

Catalog No. ICN19517391


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethylenediamine tetraacetic acid disodium salt dihydrate
White
MFCD00150037,MFCD00003541
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
372.24
372.2
Powder
6381-92-6
C10H18N2Na2O10
1 kg
OVBJJZOQPCKUOR-UHFFFAOYSA-L
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
44120005
0.9974

Chemical Identifiers

6381-92-6
372.24
OVBJJZOQPCKUOR-UHFFFAOYSA-L
44120005
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
C10H18N2Na2O10
MFCD00150037,MFCD00003541
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions